Multiple affinity removal spin cartridge HSA/IgG
Lieferant: AGILENT
5188-8825
AGLS5188-8825EA 535 EUR
AGLS5188-8825
Multiple affinity removal spin cartridge HSA/IgG
Chromatographiesäulen
The multiple affinity removal spin cartridge – Human Albumin/IgG is designed to remove two interfering high-abundant proteins from human samples (such as plasma, serum, urine or cerebrospinal fluid).
  • Compatible with standard benchtop microcentrifuges and luer-lok syringes
  • Serum load volume is 50 µl per run
  • Optimise sample recovery by utilising our rapid, single injection protocols and optimised buffers
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.
Bitte beachten For research use only. Not for use in diagnostic procedures.