MVP human total RNA
Lieferant: STRATAGENE
540025 540025
STRA540025EA 157 EUR
STRA540025 AGLS540025
MVP human total RNA
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
MVP RNA is application-ready RNA available in small economical pack sizes.
  • Eliminates tedious, time consuming RNA isolation procedures
  • Small, economically priced and convenient 25 µg pack sizes
  • Matched sets of normal and diseased tissue from the same donor
  • Well-documented donor, tissue pathology information
Extensive and rigorous quality control including many application specific assays provides assurance that the RNA is intact, full-length and nearly DNA-free.
Application: Real time RT-PCR, miRNA detection and Northern blot analysis.