MVP human total RNA
Lieferant: STRATAGENE
540025
540025
STRA540025EA
157
EUR
STRA540025
AGLS540025
MVP human total RNA
Nukleinsäure-Reagenzien
qPCR/RT-qPCR Enzyme und Kits
MVP RNA is application-ready RNA available in small economical pack sizes.
- Eliminates tedious, time consuming RNA isolation procedures
- Small, economically priced and convenient 25 µg pack sizes
- Matched sets of normal and diseased tissue from the same donor
- Well-documented donor, tissue pathology information
Extensive and rigorous quality control including many application specific assays provides assurance that the RNA is intact, full-length and nearly DNA-free.
Application: Real time RT-PCR, miRNA detection and Northern blot analysis.