Perfect Match PCR enhancers
Lieferant: AGILENT
600129
600130
AGLS600129EA
482
EUR
AGLS600129
AGLS600130
Perfect Match PCR enhancers
Nukleinsäure-Reagenzien
qPCR/RT-qPCR Enzyme und Kits
Increases specificity and yield of primary PCR amplification products.
- Minimises formation of poorly matched primer-template complexes
- Add to genomic PCR reactions to destabilise many mismatched primer-template complexes
Explore how Perfect Match PCR enhancer increases the specificity and yield of primary PCR amplification products. When added to genomic in vitro amplification reactions, it destabilises numerous mismatched primer-template complexes that would otherwise result in a heterogeneous population of amplified molecules.
Bitte beachten For research use only. Not for use in diagnostic procedures.