Taq extender PCR additive
Lieferant: AGILENT
600148
AGLS600148EA
291
EUR
AGLS600148
Taq extender PCR additive
Nukleinsäure-Reagenzien
qPCR/RT-qPCR Enzyme und Kits
Improves reliability and yield of conventional Taq-based PCR amplifications.
- Improves length, yield, and reliability of PCR
- Enhances efficiency of template extension by Taq DNA polymerase
- Improves amplification of difficult targets
Taq Extender PCR additive improves the reliability and yield of conventional Taq-based PCR amplifications. Taq Extender additive increases the efficiency of Taq DNA polymerase extension reactions during each cycle of PCR by reducing mismatch pausing, resulting in a greater percentage of completed extension reactions.
Bitte beachten For research use only. Not for use in diagnostic procedures.