Brilliant II probe MM
Lieferant: AGILENT
600804
600809
600810
600815
600818
600819
600827
AGLS600804EA
566
EUR
AGLS600804
AGLS600809
AGLS600810
AGLS600815
AGLS600818
AGLS600819
AGLS600827
Brilliant II probe MM
Nukleinsäure-Reagenzien
qPCR/RT-qPCR Enzyme und Kits
Brilliant II QPCR and QRT-PCR reagents were developed for improved sensitivity of detection, ensuring reproducible quantification even at low target concentrations.
- Greater flexibility for use with numerous different templates and targets
- Developed on the Mx QPCR Systems
- Passive reference dye to maximise performance on different instrument platforms
- Multiplex kit for improved results on multiple targets
For faster experiments with earlier Ct's and greater reproducibility, Brilliant III Ultra-Fast master mixes can be used.
Bitte beachten For research use only. Not for use in diagnostic procedures.