Brilliant II SYBR® master mix with ROX
Lieferant: AGILENT
600830
AGLS600830EA 566 EUR
AGLS600830
Brilliant II SYBR® master mix with ROX
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
Brilliant II SYBR® QPCR and QRT-PCR master mix kits with high/low Rox offer superior sensitivity for improved quantification and reproducibility.
  • Improved performance with earlier Ct detection across a wide dynamic range
  • Increased sensitivity of detection to low copy numbers
It also shows more consistent performance across a wide range of template concentrations and different targets.
Bitte beachten For research use only. Not for use in diagnostic procedures.