Brilliant III Probe MM, Agilent Technologies
Lieferant: AGILENT
600880
600881
600884
600885
732-3798EA
429
EUR
732-3798
AGLS600881
AGLS600884
AGLS600885
Brilliant III Probe MM, Agilent Technologies
Nukleinsäure-Reagenzien
qPCR/RT-qPCR Enzyme und Kits
Brilliant III QPCR and QRT-PCR master mixes increase overall sensitivity and speed through the novel, quick-activating hot start and the engineered Taq-mutant.
- Extremely fast while maintaining sensitivity
- Greater resistance to common QPCR inhibitors (whole blood or NaCl)
- Optimized fast cycling formulation ensures reliable and reproducible data with shorter run times
- Convenient pre-blended formulations compatible with any sequence-specific probe detection chemistry
Brilliant III Ultra-Fast maintains amplification efficiency, R2, dynamic range and detection sensitivity all while shortening the total cycling time by approximately 60% compared to standard cycling.
Bitte beachten For research use only. Not for use in diagnostic procedures.