qPCR NGS library quantification kit
Lieferant: AGILENT
The Agilent qPCR NGS Library quantification kit is a reliable method for quantifying NGS libraries.
- Provides researchers with an accurate and sensitive method for quantifying NGS libraries
- Validated in the SureSelect Target Enrichment Protocol for barcoding and indexing applications
- Illumina and AB SOLID library quantification kits available
The Agilent qPCR NGS Library quantification kit provides researchers an accurate and sensitive method for quantifying NGS libraries. The NGS Library Quantification kit was validated in the SureSelect Target Enrichment Protocol for barcoding and indexing applications. Illumina and AB SOLID library quantification kits available.
Accurate library quantification leads to optimal cluster densities for improved sequence efficiency and data quality. Consistent quantification across a broad range of samples, varying library insert sizes, and GC content. Quantify 84 libraries with each kit.