Fragment Analyzer System, 5300
Lieferant: AGILENT
M5311AA
AGLSM5311AAEA 82080 EUR
AGLSM5311AA
Fragment Analyzer System, 5300
Elektrophorese-Systeme Kapillarelektrophorese-Systeme
Parallel capillary electrophoresis system used for nucleic acid QC in medium to high-throughput labs.
  • Separate 48 or 96 samples in parallel with a single simple dilution step and easy instrument set up
  • Reduce sample analysis time by using batch processes features post electrophoresis to automatically generate results reports
  • Load and separate up to 288 samples for unattended and automated analysis of large numbers of samples
  • Easily adapts to changes in your workflow with flexible option like maximising your runs or speed or resolution of individual samples by choosing a shorter or longer array
  • Minimise hands on time with no daily array handling and room-temperature stable reagents
  • Change priority of runs to fit your lab’s needs
  • Use quality metrics for RNA (RQN) and genomic DNA (GQN) to remove subjective quality assessments; See clear results with the high sizing accuracy and precision along with separation resolution as good as 3 bp from fragments under 300 bp
  • Kits provide a wide dynamic range covering two orders of magnitude
  • Achieve accurate molarity calculations with reliable smear analysis
  • IQ/OQ services available for use in GMP environments
  • Desktop PC supplied with a comprehensive 3-year warranty through HP
The 5300 Fragment Analyzer system is a capillary electrophoresis instrument that can separate up to 48 or 96 samples in parallel. This system is designed to improve lab efficiency and workflow allowing you to automate the analysis of large numbers of samples and focus on results.
The 5300 Fragment Analyzer system performs DNA QC and RNA QC for a broad range of samples including, gDNA, small RNA, cfDNA, large DNA fragments, and total RNA. The diversity of sample types these systems can separate make these instruments ideal for individualised workflows, including NGS library QC and CRISPR workflows.
Bitte beachten For research use only. Not for use in diagnostic procedures.