Abscisic Acid ELISA Quantitation Kit
Lieferant: Agrisera
AS204392-5X96
AGRIAS204392-5X96EA 2220 EUR
AGRIAS204392-5X96
Abscisic Acid ELISA Quantitation Kit
Tests ELISAs
This ELISA assay utilises the principle of competitive binding to measure the concentration of hormone in plant extracts.
The ABA (abscisic acid) hormone has been pre-coated on the surface of the reaction wells. The plant extract sample, containing an unknown amount of hormone, or standards are mixed in the reaction well with a known amount of antibody to ABA. During incubation, the competitive inhibition reaction occurs between pre-coated ABA and hormone in the samples with the ABA-specific antibody.