Portable oxygen detector
Lieferant: Air Liquide
ACCSECU26
AIRLACCSECU26EA 311.02 EUR
AIRLACCSECU26
Portable oxygen detector
Gas-Spürsysteme
Oxygen detector for safety and compliance in harsh environments with extreme temperatures.
  • Maintenance-free: No sensor or battery change
  • Hibernation mode with case accessory or IntelliDox
  • Automated self-test of battery, sensor and electronics
  • Wide-angle flash, which alerts with audible and vibrating alarm
  • Automatic logging of the 35 latest events and bump test results
Standards: ATEX: CE 0539 II 1G; Ex ia IIC T4 Ga; DEMKO 14 ATEX 1356 - IECEx: Ex ia IIC T4 Ga; IECEx UL 14.0063