Cortisol ELISA kit

Lieferant: Enzo Life Sciences

ADI-900-071 ADI-901-071 ADI900071 ADI901071
ALEXADI-900-071EA 0 EUR
ALEXADI-900-071 ALEXADI-901-071 ENZOADI900071 ENZOADI901071
Cortisol ELISA kit
Tests ELISAs

The Cortisol EIA kit is a colorimetric competitive enzyme immunoassay kit for the quantitative determination of cortisol in culture supernatants, plasma, serum, saliva, and urine from any species.


  • Trusted - highly cited in peer reviewed literature
  • Reproducible results day-after-day and lot-after-lot
  • Highly sensitive, detecting low levels of cortisol in biological matrices
  • Pre-coated plates and liquid colour coded reagents save time and reduce errors


Absorbance is read at 405 nm. The broad dynamic range allows accurate measurement of cortisol levels in a variety of sample matrices.


Bestellinformation: Contains GxM IgG microtiter plate, conjugate, antibody, assay buffer, wash buffer concentrate, standard, pNpp substrate, stop solution, steroid displacement reagent.

Bestellen

TECHNISCHE DATEN

Environmentally Preferable
Reaktivität der primären Antikörper
Testtyp Competitive
Wirt
Empfindlichkeit 56,72 pg/ml (range 156 - 10000 pg/ml) 56,72 pg/ml (range 156 - 10000 pg/ml)
Lagerbedingungen 4 °C (long term) 4 °C (long term)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR