Paraformaldehyd 4% PBS 91-93
Lieferant: Thermo Fisher Scientific
Synonyme:
Polyoxymethylen, Polymethanal
J19943.K2
ALFAJ19943.K2EA
130
EUR
ALFAJ19943.K2
Paraformaldehyd 4% PBS 91-93
Paraformaldehyd
Formel:
(CH₂O)n Schmelzpunkt: 123 Flammpunkt: 70 |
MDL:
MFCD00133991 CAS-Nummer: 30525-89-4 Merck Index: 13,07096 |
Spezifikation Testergebnisse
Form | Colourless, clear solution |
Assay (unspecified) (formaldehyde) | 91 − 93% |
pH (25 °C) | 7,0 − 7,6 |
Concentration | 1XPBS (0,01M) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...