BioHazard Tray, White
Lieferant: ALPHA LABORATORIESCHEMICALS
EL2674
ALPCEL2674EA 249 EUR
ALPCEL2674
BioHazard Tray, White
Schalen/Schüsseln/Töpfe Auffangschalen und Auffangbehälter
The biohazard spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.
Lieferumfang: All trays are supplied with one free liner when purchased.