HIGH STAT PLUS ESD Jackets
Lieferant: ALSICO HIGHTECH
1017456-XS 1017456-S 1017456-M 1017456-L 1017456-XL 1017456-2XL
ALSO1017456-XSEA 82.5 EUR
ALSO1017456-XS ALSO1017456-S ALSO1017456-M ALSO1017456-L ALSO1017456-XL ALSO1017456-2XL
HIGH STAT PLUS ESD Jackets
Arbeitskleidung
Unisex ESD jackets.
  • Upright collar
  • Centre front closure with hidden zipper and press studs
  • Adjustable waistband with press studs
  • Chest pocket left with zipper
  • Chest pocket right with flap and hidden press studs
  • 2 side pockets with vertical zipper
  • Inset sleeves
  • Adjustable sleeve ends with press studs
  • Accents in royal blue
  • ESD logo
Industrial launderable.
Standards: Oeko-Tex® standard 100; IEC 61340 - 4 - 9:2016.