Cleanroom Hoods, Balloon Tops
Lieferant: ALSICO HIGHTECH
1033541-XS 1033541-S 1033541-M 1033541-L 1033541-XL 1033541-XXL 1033541-3XL 1033542-XS 1033542-S 1033542-M 1033542-L 1033542-XL 1033542-XXL 1033542-3XL
ALSO1033541-XSEA 33 EUR
ALSO1033541-XS ALSO1033541-S ALSO1033541-M ALSO1033541-L ALSO1033541-XL ALSO1033541-XXL ALSO1033541-3XL ALSO1033542-XS ALSO1033542-S ALSO1033542-M ALSO1033542-L ALSO1033542-XL ALSO1033542-XXL ALSO1033542-3XL
Cleanroom Hoods, Balloon Tops
Arbeitskleidung
These hoods are made of Alsistrong® (98% polyester and 2% carbon). Suitable together with a coverall for the highest cleanroom classes ISO 4 to 9 or GMP classes A to D.
  • Balloon top
  • Concealed zip front fastening with studs
  • Studs to attach face mask
  • Side openings for cleanroom goggles
  • Strap with buckle at the back
Usable in ISO 4 to 9.
Standards: IEST-RP-CC003.5.