Cleanroom lab coats
Lieferant: ALSICO HIGHTECH
1037468-XS 1037468-S 1037468-M 1037468-L 1037468-XL 1037468-XXL 1037468-3XL 1037470-XS 1037470-S 1037470-M 1037470-L 1037470-XL 1037470-XXL 1037470-3XL
ALSO1037468-XSEA 59.2 EUR
ALSO1037468-XS ALSO1037468-S ALSO1037468-M ALSO1037468-L ALSO1037468-XL ALSO1037468-XXL ALSO1037468-3XL ALSO1037470-XS ALSO1037470-S ALSO1037470-M ALSO1037470-L ALSO1037470-XL ALSO1037470-XXL ALSO1037470-3XL
Cleanroom lab coats
Arbeitskleidung
This cleanroom coat is made of Alsistrong® (98% polyester and 2% carbon). Suitable for C and D classes or ISO 7 to 9 cleanrooms. Can also be used for clean areas or as lab coat.
  • Stand collar with two studs
  • Concealed zip front fastening
  • Raglan sleeves with antistatic cuffs
  • Studs for name tag on left chest
  • Marker fabric
  • No pockets
Standards: IEST-RP-CC003.4