Lab Coats, White
Lieferant: ALSICO HIGHTECH
1039364W-L 1039364W-M 1039364W-S 1039364W-XL 1039364W-XS 1039364W-XXL 1039364W-3XL
ALSO1039364W-LEA 0 EUR
ALSO1039364W-L ALSO1039364W-M ALSO1039364W-S ALSO1039364W-XL ALSO1039364W-XS ALSO1039364W-XXL ALSO1039364W-3XL
Lab Coats, White
Arbeitskleidung
This cleanroom coat is made of Alsistrong® (98% polyester and 2% carbon).
  • Hidden two-way front zipper
  • 1 left chest pocket
  • 2 side pockets
  • Marker fabric
  • High comfort logo
  • 118 g/m2
All lab coats feature an upstanding collar with two studs for adjustment, and raglan sleeves with anti-static cuffs.