Digital thermometer, Maxi Pen
Lieferant: Amarell
E905190
AMARE905190EA 32.92 EUR
AMARE905190
Digital thermometer, Maxi Pen
Thermometer Digitalthermometer
Resistant and intensive thermometer for fast temperature indication. With fast-responding stainless steel penetration probe (L×Ø: 125×3,5 mm).
  • 'HOLD' feature
  • Automatic power-off after 10 min.
  • °C / °F switchable
  • Waterproof casing