Digital thermometer, wireless
Lieferant: Amarell
E907600
AMARE907600EA 41.91 EUR
AMARE907600
Digital thermometer, wireless
Thermometer Digitalthermometer
Wireless multi-channel thermometer as desktop model. Three channels connectable on one base terminal.
  • Switchable from °C to F
  • Alert option
Lieferumfang: Includes a remote sensor.