Digital timer
Lieferant: Amarell
E920630
AMARE920630EA 19.86 EUR
AMARE920630
Digital timer
Timer Timer, digital
Count down/count up timer with clock and stopwatch.
  • Timing capacity: 24 hours
  • Loud alarm signal
  • Memory function
  • With magnet, standing and holding device
Lieferumfang: Supplied with round cell LR44/A76.