Anti-ASK1 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A12601-100
ANTIA12601-100EA 415 EUR
ANTIA12601-100
Anti-ASK1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to ASK1 for WB with samples derived from human and mouse.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:200

Type: Primary
Antigen: ASK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR