Human Mycoplasma Pneumoniae IgG ELISA Kit
Lieferant: ANTIBODIES.COM
Human Mycoplasma pneumoniae IgG ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Mycoplasma pneumoniae IgG in serum, plasma, tissue homogenates, and other biological fluids.