Anti-Iba1 Mouse Recombinant Antibody [clone: rAIF1/1909]
Lieferant: ANTIBODIES.COM
A248418-100
ANTIA248418-100EA 520 EUR
ANTIA248418-100
Anti-Iba1 Mouse Recombinant Antibody [clone: rAIF1/1909]
Antikörper
Recombinant mouse monoclonal [rAIF1/1909] antibody to Iba1 for IHC-P with samples derived from human.
  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Iba1
Clonality: Recombinant
Clone: rAIF1/1909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human