Anti-Iba1 Mouse Monoclonal Antibody [clone: AIF1/2493]

Lieferant: ANTIBODIES.COM
A248419-100
ANTIA248419-100EA 520 EUR
ANTIA248419-100
Anti-Iba1 Mouse Monoclonal Antibody [clone: AIF1/2493]
Antikörper
Mouse monoclonal [AIF1/2493] antibody to Iba1 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Iba1
Clonality: Monoclonal
Clone: AIF1/2493
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR