Anti-IDH1 Mouse Monoclonal Antibody [clone: IDH1/1152]

Lieferant: ANTIBODIES.COM
A248916-100
ANTIA248916-100EA 520 EUR
ANTIA248916-100
Anti-IDH1 Mouse Monoclonal Antibody [clone: IDH1/1152]
Antikörper
Mouse monoclonal [IDH1/1152] antibody to IDH1 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: IDH1
Clonality: Monoclonal
Clone: IDH1/1152
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR