Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: LLC/1738]

Lieferant: ANTIBODIES.COM
A248998-100
ANTIA248998-100EA 520 EUR
ANTIA248998-100
Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: LLC/1738]
Antikörper
Mouse monoclonal [LLC/1738] antibody to Lambda Light Chain for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Lambda Light Chain
Clonality: Monoclonal
Clone: LLC/1738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR