Anti-Lambda Light Chain Rabbit Recombinant Antibody [clone: LLC/3778R]
Lieferant: ANTIBODIES.COM
A249000-100
ANTIA249000-100EA 520 EUR
ANTIA249000-100
Anti-Lambda Light Chain Rabbit Recombinant Antibody [clone: LLC/3778R]
Antikörper
Recombinant rabbit monoclonal [LLC/3778R] antibody to Lambda Light Chain for IHC-P with samples derived from human.
  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Lambda Light Chain
Clonality: Recombinant
Clone: LLC/3778R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human