Anti-PAX6 Mouse Monoclonal Antibody [clone: SPM612]

Lieferant: ANTIBODIES.COM
A249568-100
ANTIA249568-100EA 520 EUR
ANTIA249568-100
Anti-PAX6 Mouse Monoclonal Antibody [clone: SPM612]
Antikörper
Mouse monoclonal [SPM612] antibody to PAX6 for flow cytometry, IF and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IF, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: PAX6
Clonality: Monoclonal
Clone: SPM612
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR