Anti-PAX7 Mouse Monoclonal Antibody [clone: PAX7/1187]

Lieferant: ANTIBODIES.COM
A249570-100
ANTIA249570-100EA 520 EUR
ANTIA249570-100
Anti-PAX7 Mouse Monoclonal Antibody [clone: PAX7/1187]
Antikörper
Mouse monoclonal [PAX7/1187] antibody to PAX7 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: PAX7
Clonality: Monoclonal
Clone: PAX7/1187
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR