Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: LLC/1738]
Lieferant: ANTIBODIES.COM
A252178-100
ANTIA252178-100EA 520 EUR
ANTIA252178-100
Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: LLC/1738]
Antikörper
Mouse monoclonal [LLC/1738] antibody to Lambda Light Chain for IHC-P with samples derived from Human.
  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Lambda Light Chain
Clonality: Monoclonal
Clone: LLC/1738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human