Anti-Pig IgG H&L Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A294504-1
ANTIA294504-1EA 160 EUR
ANTIA294504-1
Anti-Pig IgG H&L Rabbit Polyclonal Antibody
Antikörper
Rabbit anti-pig IgG H&L secondary antibody for WB, IHC, ICC and ELISA with samples derived from pig.

  • Validated applications: WB, IHC, ICC, ELISA
  • Recommended dilutions: ELISA: 1:1000 to 1:30000; ELISA (capture antibody): 1:100 to 1:500, ICC: 1:200 to 1:2000, IHC: 1:200 to 1:2000, WB: 1:1000 to 1:30000

Type: Secondary
Antigen: Pig IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR