Human UBC3B ELISA Kit
Lieferant: ANTIBODIES.COM
A303059-96
ANTIA303059-96EA 630 EUR
ANTIA303059-96
Human UBC3B ELISA Kit
Tests ELISAs
Human UBC3B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human UBC3B in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready to use ELISA kit
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human UBC3B ELISA kit (A303059) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human UBC3B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for UBC3B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the UBC3B present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-UBC3B Antibody, which binds the captured UBC3B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of UBC3B captured in each well. The concentration of UBC3B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Bestellinformation: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.