Anti-PKC gamma Rabbit Monoclonal Antibody [clone: ARC1637]

Lieferant: ANTIBODIES.COM
A305980-100
ANTIA305980-100EA 525 EUR
ANTIA305980-100
Anti-PKC gamma Rabbit Monoclonal Antibody [clone: ARC1637]
Antikörper
Rabbit monoclonal [ARC1637] antibody to PKC gamma for WB with samples derived from Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: PKC gamma
Clonality: Monoclonal
Clone: ARC1637
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR