Anti-POLD1 Rabbit Monoclonal Antibody [clone: ARC0930]
Lieferant: ANTIBODIES.COM
A306013-100
ANTIA306013-100EA 525 EUR
ANTIA306013-100
Anti-POLD1 Rabbit Monoclonal Antibody [clone: ARC0930]
Antikörper
Rabbit monoclonal [ARC0930] antibody to POLD1 for WB with samples derived from Human, Mouse and Rat.
  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: POLD1
Clonality: Monoclonal
Clone: ARC0930
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat