Anti-ELK1 Rabbit Monoclonal Antibody [clone: ARC0339]

Lieferant: ANTIBODIES.COM
A306016-100
ANTIA306016-100EA 525 EUR
ANTIA306016-100
Anti-ELK1 Rabbit Monoclonal Antibody [clone: ARC0339]
Antikörper
Rabbit monoclonal [ARC0339] antibody to ELK1 for WB and IP with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IP
  • Recommended Dilutions: WB: 1:500 to 1:2000, IP: 1:500 to 1:1000

Type: Primary
Antigen: ELK1
Clonality: Monoclonal
Clone: ARC0339
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR