Anti-Epsin 1 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A306760-100
ANTIA306760-100EA 415 EUR
ANTIA306760-100
Anti-Epsin 1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to Epsin 1 for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type: Primary
Antigen: Epsin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR