Anti-Iba1 Rabbit Monoclonal Antibody [clone: ARC2301]

Lieferant: ANTIBODIES.COM
A307823-100
ANTIA307823-100EA 525 EUR
ANTIA307823-100
Anti-Iba1 Rabbit Monoclonal Antibody [clone: ARC2301]
Antikörper
Rabbit monoclonal [ARC2301] antibody to Iba1 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: Iba1
Clonality: Monoclonal
Clone: ARC2301
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR