Human SPARC ELISA Kit

Lieferant: ANTIBODIES.COM

A310999-96
ANTIA310999-96EA 545 EUR
ANTIA310999-96
Human SPARC ELISA Kit
Tests ELISAs

Human SPARC ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SPARC in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 250 to 4000 ng/ml
  • Sensitivity: 9,48 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%


Human SPARC ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SPARC in An antibody specific for SPARC has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SPARC present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-SPARC antibody, which also binds the SPARC present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SPARC antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SPARC antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of SPARC captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of SPARC in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Bestellinformation: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR