Human Tec ELISA Kit
Lieferant: ANTIBODIES.COM
Human Tec ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Tec in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 75 to 1200 ng/L
- Sensitivity: 4,91 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Tec ELISA kit (A313619) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Tec in An antibody specific for Tec has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Tec present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Tec antibody, which also binds the Tec present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Tec antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Tec antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Tec captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Tec in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Bestellinformation: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...