Anti-Iba1 Rabbit Recombinant Antibody [clone: AIF1/8970R]

Lieferant: ANTIBODIES.COM
A315121-100
ANTIA315121-100EA 680 EUR
ANTIA315121-100
Anti-Iba1 Rabbit Recombinant Antibody [clone: AIF1/8970R]
Antikörper
Recombinant rabbit monoclonal [AIF1/8970R] antibody to Iba1 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Iba1
Clonality: Recombinant
Clone: AIF1/8970R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR