Anti-IDH1 Rabbit Recombinant Antibody [clone: IDH1.R132H/7328R]
Lieferant: ANTIBODIES.COM
A316514-100
ANTIA316514-100EA 680 EUR
ANTIA316514-100
Anti-IDH1 Rabbit Recombinant Antibody [clone: IDH1.R132H/7328R]
Antikörper
Recombinant rabbit monoclonal [IDH1.R132H/7328R] antibody to IDH1 for IHC-P with samples derived from human.
  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: IDH1
Clonality: Recombinant
Clone: IDH1.R132H/7328R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human