Anti-IDH1 Rabbit Recombinant Antibody [clone: IDH1.R132H/8205R]

Lieferant: ANTIBODIES.COM
A316515-100
ANTIA316515-100EA 680 EUR
ANTIA316515-100
Anti-IDH1 Rabbit Recombinant Antibody [clone: IDH1.R132H/8205R]
Antikörper
Recombinant rabbit monoclonal [IDH1.R132H/8205R] antibody to IDH1 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: IDH1
Clonality: Recombinant
Clone: IDH1.R132H/8205R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR