Mouse beta 2 Defensin/BD-2 ELISA Kit

Lieferant: ANTIBODIES.COM

A76435-96
ANTIA76435-96EA 600 EUR
ANTIA76435-96
Mouse beta 2 Defensin/BD-2 ELISA Kit
Tests ELISAs

Mouse beta 2 Defensin/BD-2 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse beta 2 Defensin/BD-2 in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready to use ELISA kit
  • Detection range: 62,5 to 4000 pg/ml
  • Sensitivity: 37,5 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%


Mouse beta 2 Defensin/BD-2 ELISA kit (A76435) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse beta 2 Defensin/BD-2 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with beta 2 Defensin/BD-2 antigen. During the incubation, beta 2 Defensin/BD-2 present in the samples or standards competes with the fixed amount of immobilised beta 2 Defensin/BD-2 for binding sites on the Biotinylated Anti-beta 2 Defensin/BD-2 Antibody. The more beta 2 Defensin/BD-2 present in a sample or standard, the less Biotinylated Anti-beta 2 Defensin/BD-2 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-beta 2 Defensin/BD-2 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of beta 2 Defensin/BD-2 present in each sample or standard. The concentration of beta 2 Defensin/BD-2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Bestellinformation: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR