Human Chymotrypsin ELISA Kit

Lieferant: ANTIBODIES.COM

A76479-96
ANTIA76479-96EA 600 EUR
ANTIA76479-96
Human Chymotrypsin ELISA Kit
Tests ELISAs

Human Chymotrypsin ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Chymotrypsin in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready to use ELISA kit
  • Detection range: 3,125 to 200 ng/ml
  • Sensitivity: 1,875 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%


Human Chymotrypsin ELISA kit (A76479) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Chymotrypsin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Chymotrypsin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Chymotrypsin present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Chymotrypsin Antibody, which binds the captured Chymotrypsin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Chymotrypsin captured in each well. The concentration of Chymotrypsin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Bestellinformation: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR