Mouse Antidiuretic Hormone ELISA Kit

Lieferant: ANTIBODIES.COM

A77640-96
ANTIA77640-96EA 600 EUR
ANTIA77640-96
Mouse Antidiuretic Hormone ELISA Kit
Tests ELISAs

Mouse Antidiuretic Hormone ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Antidiuretic Hormone in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready to use ELISA kit
  • Detection range: 15,625 to 1000 pg/ml
  • Sensitivity: 9,375 pg/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%


Mouse Antidiuretic Hormone employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Antidiuretic Hormone in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Antidiuretic Hormone has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Antidiuretic Hormone present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Antidiuretic Hormone Antibody, which binds the captured Antidiuretic Hormone present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Antidiuretic Hormone captured in each well. The concentration of Antidiuretic Hormone can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Bestellinformation: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR