Rat Alcohol Dehydrogenase 1 ELISA Kit

Lieferant: ANTIBODIES.COM

A79875-96
ANTIA79875-96EA 600 EUR
ANTIA79875-96
Rat Alcohol Dehydrogenase 1 ELISA Kit
Tests ELISAs

Rat Alcohol Dehydrogenase 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Alcohol Dehydrogenase 1 in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready-to-use ELISA Kit
  • Detection range: 0,625 to 40 ng/ml
  • Sensitivity: 0,375 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%


Rat Alcohol Dehydrogenase 1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Alcohol Dehydrogenase 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Alcohol Dehydrogenase 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Alcohol Dehydrogenase 1 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Alcohol Dehydrogenase 1 Antibody, which binds the captured Alcohol Dehydrogenase 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Alcohol Dehydrogenase 1 captured in each well. The concentration of Alcohol Dehydrogenase 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Bestellinformation: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR