Rat Sonic Hedgehog ELISA Kit

Lieferant: ANTIBODIES.COM

A80364-96
ANTIA80364-96EA 600 EUR
ANTIA80364-96
Rat Sonic Hedgehog ELISA Kit
Tests ELISAs

Rat Sonic Hedgehog ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Sonic Hedgehog in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready-to-use ELISA Kit
  • Detection range: 0,156 to 10 ng/ml
  • Sensitivity: 0,094 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%


Rat Sonic Hedgehog ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Sonic Hedgehog in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Sonic Hedgehog has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Sonic Hedgehog present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Sonic Hedgehog Antibody, which binds the captured Sonic Hedgehog present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Sonic Hedgehog captured in each well. The concentration of Sonic Hedgehog can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Bestellinformation: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR