Anti-IDH1 Goat Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A84905-100
ANTIA84905-100EA 475 EUR
ANTIA84905-100
Anti-IDH1 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody to IDH1 for ELISA and WB with samples derived from human and mouse.

  • Validated applications: ELISA, W

Type: Primary
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR