Anti-IDH1 Goat Polyclonal Antibody
Lieferant: ANTIBODIES.COM
A84906-100
ANTIA84906-100EA 475 EUR
ANTIA84906-100
Anti-IDH1 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody to IDH1 for ELISA and WB with samples derived from human, mouse and rat.
  • Validated applications: ELISA, W
Type: Primary
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat